missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RAVER1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA541934
This item is not returnable.
View return policy
Description
Peptide sequence: VTHRPPLSPK SGAEVEAGDA AERRAPEEEL PPLDPEEIRK RLEHTERQFR Sequence homology: Cow: 86%; Dog: 79%; Guinea Pig: 79%; Horse: 86%; Human: 100%; Rabbit: 86%.
RAVER1 gene ontology annotations related to this gene include RNA binding.
Specifications
| RAVER1 | |
| Polyclonal | |
| Unconjugated | |
| RAVER1 | |
| 1300006N24Rik; AA050359; Kiaa1978; LOW QUALITY PROTEIN: ribonucleoprotein PTB-binding 1; mKIAA1978; Protein raver-1; PTB-binding 1; RAVER1; Ribonucleoprotein; Ribonucleoprotein PTB-binding 1; ribonucleoprotein, PTB binding 1; ribonucleoprotein, PTB-binding 1 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 125950 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8IY67 | |
| RAVER1 | |
| Synthetic peptide directed towards the N-terminal of human RAVER1 (aa 7-56). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction