missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SPASKTTLHK TGIASSILDA ISQGSESQHA LEKCIAYSPP FGDYAQVSSL RNEKRGSSYL ISAPEGGSVE CVDQHSQGTG Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
RAVER2 may bind single-stranded nucleic acids.
Specifications
Specifications
| Antigen | RAVER2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.05 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | RAVER2 |
| Gene Accession No. | Q9HCJ3 |
| Gene Alias | A430091O22Rik; KIAA1579; Protein raver-2; RAVER2; Ribonucleoprotein PTB-binding 2; ribonucleoprotein, PTB binding 2; ribonucleoprotein, PTB-binding 2 |
| Gene Symbols | RAVER2 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
