missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAX Monoclonal Antibody (OTI1C5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811003S
This item is not returnable.
View return policy
Description
RAX Monoclonal antibody specifically detects RAX in Human samples. It is validated for Western BlotSpecifications
| RAX | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O14901 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 8462 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI1C5 | |
| Unconjugated | |
| KLF11 | |
| 7530406A22Rik; E130303K03Rik; ey1; ey-1; eyeless 1; LOW QUALITY PROTEIN: retinal homeobox protein Rx; MCOP3; RAX; retina and anterior neural fold homeobox; retina and anterior neural fold homeobox protein; Retinal homeobox protein Rx; Rx; Rx/rax homeoprotein | |
| Human recombinant protein fragment corresponding to amino acids 1-274 of human KLF11 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction