missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RAX, Mouse anti-Human, Clone: 3B3, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00030062-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: KEPWEARPSPGLPVGPATGEAKLSEEEQPKKKHRRNRTTFTTYQLHELERAFEKSHYPDVYSREELAGKVNLPEVRVQVWFQNRRAKWRRQEKLEVSSMKLQDSpecifications
| RAX | |
| Monoclonal | |
| Unconjugated | |
| retina and anterior neural fold homeobox | |
| MCOP3/RX | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Immunofluorescence, Western Blot | |
| 3B3 | |
| In 1x PBS, pH 7.4 | |
| NM_013435 | |
| RAX | |
| RAX (NP_038463, 104 a.a. ∼ 206 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 30062 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction