missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Antibody (RB1/2313R), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-08940CL1
This item is not returnable.
View return policy
Description
RB1 Monoclonal antibody specifically detects RB1 in Human,Mouse samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RB1 | |
| Monoclonal | |
| CoraFluor 1 | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| Recombinant human full-length RB1 protein (Uniprot: P06400) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 5925 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| Immunohistochemistry (Paraffin) | |
| RB1/2313R | |
| PBS | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction