missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Antibody (RB1/7080R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 310.00 - € 564.00
Specifications
| Antigen | RB1 |
|---|---|
| Clone | RB1/7080R |
| Concentration | 0.2 mg/ml |
| Dilution | Immunohistochemistry-Paraffin 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621664
|
Novus Biologicals
NBP3-20404-20ug |
20 μg |
€ 310.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18674804
|
Novus Biologicals
NBP3-20404-100ug |
100 μg |
€ 564.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| RB1 | |
| 0.2 mg/ml | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cell Cycle and Replication, Phospho Specific, Transcription Factors and Regulators, Tumor Suppressors | |
| 10mM PBS with 0.05% BSA | |
| 5925 | |
| IgG κ | |
| Protein A or G purified |
| RB1/7080R | |
| Immunohistochemistry-Paraffin 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| Recombinant human full-length RB1 protein | |
| Primary | |
| Store at 4&drg;C. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title