missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Antibody (SPM353), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54477JF646
This item is not returnable.
View return policy
Description
RB1 Monoclonal antibody specifically detects RB1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin)
Specifications
| RB1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| OSRC, osteosarcoma, p105-Rb, pp110, pRb, Rb, retinoblastoma 1, retinoblastoma suspectibility protein, retinoblastoma-associated protein | |
| Recombinant human RB1 protein fragment of 283 amino acid residues. (Uniprot: P06400) | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication, Transcription Factors and Regulators, Tumor Suppressors | |
| 5925 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| SPM353 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction