missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RB1 Monoclonal Antibody (OTI10H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA805671
This item is not returnable.
View return policy
Description
Rb is a tumor suppressor nuclear phosphoprotein capable of binding to DNA. It is phosphorylated on serine and threonine, but not on tyrosine residues. It forms a complex with SV40 large T antigen, adenovirus E1A, and human papilloma virus-16 E. Rb protein may act by regulating transcription and loss of its function leads to uncontrolled cell growth. Aberrations in the RB gene have been implicated in cancers of breast, colon, prostate, kidney, nasopharynx, and leukemia.Specifications
| RB1 | |
| Monoclonal | |
| Unconjugated | |
| RB1 | |
| OSRC, PPP1R130, RB, p105-Rb, pRb, pp110 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 5925 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI10H9 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06400 | |
| RB1 | |
| Human recombit protein fragment corresponding to amino acids 309-590 of human RB1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction