Learn More
RB1CC1 Mouse, anti-Human, Clone: 1G8C8, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 68564-1-IG-20UL
 View more versions of this product
Product Code. 18036695
  • € 145.00 / 20µL
Description
The RB1CC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RB1CC1. This antibody recognizes human, mouse, rat antigen. The RB1CC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
RB1CC1
1G8C8
CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC
20 μL
9821
Store at -20°C. Stable for one year after shipment.
Liquid
Western Blot, ELISA
1000 μg/mL
Mouse
Primary
Human, Mouse, Rat
150 kDa