Learn More
RB1CC1 Mouse, anti-Human, Clone: 1G8C8, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68564-1-IG-20UL
Description
The RB1CC1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human RB1CC1. This antibody recognizes human, mouse, rat antigen. The RB1CC1 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| RB1CC1 | |
| 1G8C8 | |
| CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC | |
| 20 μL | |
| 9821 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat | |
| 150 kDa | |
| IgG1 |