Learn More
RB1CC1 Rabbit, anti-Human, CoraLite Plus 488, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
€ 145.00 - € 450.00
Specifications
| Antigen | RB1CC1 |
|---|---|
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Conjugate | CoraLite Plus 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18008175
|
Proteintech
CL488-17250-100UL |
100 μL |
€ 450.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18095135
|
Proteintech
CL488-17250-20UL |
20 μL |
€ 145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The RB1CC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RB1CC1. This antibody recognizes human, mouse, rat antigen. The RB1CC1 antibody has been validated for the following applications: WB analysis. This antibody is coralite™ plus 488.
Specifications
| RB1CC1 | |
| Western Blot | |
| Liquid | |
| Human, Mouse, Rat | |
| CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC | |
| Primary |
| 1000 μg/mL | |
| CoraLite Plus 488 | |
| Rabbit | |
| 9821 | |
| IgG | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.