Learn More
RB1CC1 Rabbit, anti-Human, CoraLite Plus 750, Polyclonal, Proteintech
Product Code. 18094735 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
100 μL
20 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18094735 100 μL 100µL
18004735 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 18094735 Supplier Proteintech Supplier No. CL75017250100UL
Rabbit Polyclonal Antibody
The RB1CC1 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human RB1CC1. This antibody recognizes human, mouse, rat antigen. The RB1CC1 antibody has been validated for the following applications: FC (Intra) analysis. This antibody is coralite™ plus 750.
Specifications
Antigen RB1CC1
Applications Flow Cytometry (Intracellular Staining)
Concentration 1000 μg/mL
Conjugate CoraLite Plus 750
Gene Symbols CC1, DRAGOU14, FIP200, KIAA0203, RB1 inducible coiled coil 1, RB1CC1, RBICC
Host Species Rabbit
Quantity 100 μL
Primary or Secondary Primary
Gene ID (Entrez) 9821
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid exposure to light. Stable for one year after shipment.
Antibody Molecular Weight 150 kDa
Form Liquid
Isotype IgG