missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP 4/7 Antibody (968721), DyLight 650, R&D Systems™
Mouse Monoclonal Antibody
Brand: Novus Biologicals MAB9457C
This item is not returnable.
View return policy
Description
RBBP 4/7 Monoclonal specifically detects RBBP 4/7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RBBP 4/7 | |
| Monoclonal | |
| DyLight 650 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| E. coli-derived recombinant human RBBP-4 | |
| 100 μL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 968721 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction