missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RbBP5 Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB600-252AF488
This item is not returnable.
View return policy
Description
RbBP5 Polyclonal antibody specifically detects RbBP5 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| RbBP5 | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| RBQ3RBBP-5, retinoblastoma binding protein 5, Retinoblastoma-binding protein RBQ-3, Set1c WD40 repeat protein, homolog, SWD1 | |
| The immunogen recognized by this antibody maps to a region between residue 500 and the C-terminus (residue 538) of human retinoblastoma binding protein 5 using the numbering given in entry NP_005048.2 (GeneID 5929). | |
| 0.1 mL | |
| Cancer, Chromatin Modifiers | |
| 5929 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction