missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RbBP5 Antibody (PSH0-15), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-32895
This item is not returnable.
View return policy
Description
RbBP5 Monoclonal antibody specifically detects RbBP5 in Human,Mouse,Rat samples. It is validated for Western Blot
Specifications
| RbBP5 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 | |
| RBQ3RBBP-5, retinoblastoma binding protein 5, Retinoblastoma-binding protein RBQ-3, Set1c WD40 repeat protein, homolog, SWD1 | |
| Synthetic peptide within human RbBP5 aa 431-480/ 538. (Uniprot: Q15291) | |
| 100 μL | |
| Cancer, Chromatin Modifiers | |
| 5929 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| PSH0-15 | |
| Unconjugated | |
| PBS (pH7.4), 0.1% BSA and 40% Glycerol | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction