missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBBP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA563522
This item is not returnable.
View return policy
Description
Immunogen sequence: HGTRGELLLD VAWHPVRPII ASISSGVVSI WAQNQVENWS AFAPDFKELD ENVEYEERES EFDIEDEDKS EPEQTGADAA EDEEVDVTSV DPIAAFCSSD EELEDSKALL YLPIA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifications
| RBBP5 | |
| Polyclonal | |
| Unconjugated | |
| Rbbp5 | |
| 4933411J24Rik; C330016J05; RB binding protein 5, histone lysine methyltransferase complex subunit; RBBP 5; Rbbp5; RBBP-5; RBQ 3; RBQ3; retinoblastoma binding protein 5; retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1; SWD1, Set1c WD40 repeat protein, homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5929 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q15291 | |
| Rbbp5 | |
| Recombinant protein fragment of Human RBBP5 (Product #RP-91619) | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction