Learn More
RBBP8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12624-1-AP-150UL
This item is not returnable.
View return policy
Description
CtIP, a 125 kD protein, was originally found interacting with a transcription repressor, CtBP, through the PLDLS motif (CtBP-Interacting Protein) thus suggested a role in transcription. Studies have shown that CtIP also interacts with BRCA1 protein through the c-terminus BRCT domains also suggested that CtIP is a potential tumor suppressor. This CtIP-BRCA1 interaction can be disruppted by DNA damaging agents including UV or gamma-irradiation. Li et al (Nature 406, 210 - 215 (2000)) have shown that ATM phosphorylates CtIP at serine residues 664 and 745, and mutation of these sites abrogates the dissociation of BRCA1 from CtIP, resulting in persistent repression of BRCA1-dependent induction of GADD45 upon ionizing radiation.Specifications
| RBBP8 | |
| Polyclonal | |
| Unconjugated | |
| RBBP8 | |
| CtBP interacting protein, CTIP, DNA endonuclease RBBP8, RBBP 8, RBBP8, RIM, SAE2 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 225182, 291787, 5932 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B1WC58, Q80YR6, Q99708 | |
| Rbbp8 | |
| RBBP8 Fusion Protein Ag3314 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |