missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP9 Antibody (OTI4G3), PE/Cy7, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-73828PECY7
This item is not returnable.
View return policy
Description
RBBP9 Monoclonal specifically detects RBBP9 in Human, Mouse, Rat samples. It is validated for Flow Cytometry.
Specifications
| RBBP9 | |
| Monoclonal | |
| PE-Cyanine7 | |
| PBS with 0.05% Sodium Azide | |
| RBBP9 | |
| Full length recombinant protein of human RBBP9 (NP_006597) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
| Flow Cytometry | |
| OTI4G3 | |
| Flow Cytometry | |
| B5T-overexpressed gene protein, Bog, BOGB5T overexpressed gene protein, EC 3.-, Protein BOG, putative hydrolase RBBP9, RBBP10, RBBP-10, RBBP-9, retinoblastoma binding protein 9, Retinoblastoma-binding protein 10, Retinoblastoma-binding protein 9MGC9236, retinoma-binding protein 9 | |
| Mouse | |
| Affinity Purified | |
| Signal Transduction | |
| 10741.0 | |
| Store at 4°C in the dark. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction