Learn More
RBBP9 Monoclonal Antibody (OTI1C10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501677S
This item is not returnable.
View return policy
Description
RBBP9 Monoclonal antibody specifically detects RBBP9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| RBBP9 | |
| Monoclonal | |
| 0.29 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75884 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 10741 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI1C10 | |
| Unconjugated | |
| RBBP9 | |
| B5T overexpressed gene protein; B5T-overexpressed gene protein; BOG; im:7247963; im:7247963 protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RB binding protein 9, serine hydrolase; RBBP10; RBBP-10; Rbbp9; RBBP-9; retinoblastoma binding protein 9; retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; serine hydrolase RBBP9; serine hydrolase RBBP9; putative hydrolase RBBP9; si:ch211-258l4.7; zgc:152807 | |
| Full length human recombinant protein of human RBBP9 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |