missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP9 Monoclonal Antibody (OTI2G2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501676
This item is not returnable.
View return policy
Description
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.Specifications
| RBBP9 | |
| Monoclonal | |
| Unconjugated | |
| RBBP9 | |
| BOG, RBBP10 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10741 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2G2 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75884 | |
| RBBP9 | |
| Full length human recombit protein of human RBBP9 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction