missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBBP9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 12230-2-AP-20UL
This item is not returnable.
View return policy
Description
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.Specifications
| RBBP9 | |
| Polyclonal | |
| Unconjugated | |
| RBBP9 | |
| B5T overexpressed gene protein, BOG, Protein BOG, Putative hydrolase RBBP9, RBBP 10, RBBP 9, RBBP10, RBBP9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10741, 26450, 29459 | |
| -20°C | |
| Liquid |
| Western Blot, Immunohistochemistry (Paraffin), Immunoprecipitation, Flow Cytometry | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| O75884, O88350, O88851 | |
| Rbbp9 | |
| RBBP9 Fusion Protein Ag2868 | |
| 20 μL | |
| Primary | |
| Human, Rat, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction