missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBEL1 Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-12807JF646
This item is not returnable.
View return policy
Description
RBEL1 Polyclonal antibody specifically detects RBEL1 in Human samples. It is validated for Western Blot
Specifications
| RBEL1 | |
| Polyclonal | |
| Western Blot | |
| bA216L13.9, chromosome 9 open reading frame 86, FLJ10101, FLJ13045, KIAA1984, Parf, Partner of ARF, pp8875, putative GTP-binding protein Parf, Rab-like GTP-binding protein 1, Rab-like GTP-binding protein 1C, Rab-like protein 1, RBEL1 | |
| The immunogen this antibody was made to, maps to a region between residue 525 and 575 of human Rab-like Protein 1 using the numbering given in entry NP_078994.3 (GeneID 55684). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 55684 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction