missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPAS1 Monoclonal Antibody (OTI5B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809632
This item is not returnable.
View return policy
Description
NPAS1 is a member of the basic helix-loop-helix (bHLH)-PAS family of transcription factors. Studies of a related mouse gene suggest that it functions in neurons. The exact function of this gene is unclear, but it may play protective or modulatory roles during late embryogenesis and postnatal development.Specifications
| NPAS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q99742 | |
| NPAS1 | |
| Human recombit protein fragment corresponding to amino acids 319-522 of human NPAS1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5B2 | |
| Unconjugated | |
| NPAS1 | |
| MOP5, PASD5, bHLHe11 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4861 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction