missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX1 Monoclonal Antibody (OTI1F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803144S
This item is not returnable.
View return policy
Description
RBFOX1 Monoclonal antibody specifically detects RBFOX1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| RBFOX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NWB1 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 54715 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F7 | |
| Unconjugated | |
| Rbfox1 | |
| 2BP1; A FOX-1; A2BP; A2BP1; ataxin 2 binding protein 1; ataxin 2-binding protein 1; ataxin-2-binding protein 1; FOX1; fox-1; fox-1 homolog A; fox-1-like RNA-binding protein 1; hexaribonucleotide binding protein 1; hexaribonucleotide binding protein 1 isoform alpha; hexaribonucleotide binding protein 1 isoform beta; hexaribonucleotide-binding protein 1; HRNBP1; MNCb-3035; RBFOX1; RNA binding fox-1 homolog 1; RNA binding protein fox-1 homolog 1; RNA binding protein, fox-1 homolog (C. elegans) 1; RNA binding protein, fox-1 homolog 1 | |
| Human recombinant protein fragment corresponding to amino acids 1-210 of human RBFOX1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction