missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPAS1 Monoclonal Antibody (OTI5B2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809632S
This item is not returnable.
View return policy
Description
NPAS1 Monoclonal antibody specifically detects NPAS1 in Human samples. It is validated for Western BlotSpecifications
| NPAS1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P09496 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1211 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI5B2 | |
| Unconjugated | |
| CLTA | |
| basic-helix-loop-helix-PAS protein MOP5; BHLHE11; Class E basic helix-loop-helix protein 11; member of PAS protein 5; member of PAS superfamily 5; MOP5; neuronal PAS domain protein 1; neuronal PAS domain-containing protein 1; Neuronal PAS1; Npas1; PAS domain-containing protein 5; PASD5 | |
| Full length human recombinant protein of human CLTA produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?