missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX1 Monoclonal Antibody (OTI1F7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA803144
This item is not returnable.
View return policy
Description
Ataxin-2 binding protein 1 has an RNP motif that is highly conserved among RNA-binding proteins. This protein binds to the C-terminus of ataxin-2 and may contribute to the restricted pathology of spinocerebellar ataxia type 2 (SCA2). Ataxin-2 is the gene product of the SCA2 gene which causes familial neurodegenerative diseases. Ataxin-2 binding protein 1 and ataxin-2 are both localized in the trans-Golgi network. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene. Additional transcript variants have been found but their full length nature has not been determined.Specifications
| RBFOX1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NWB1 | |
| RBFOX1 | |
| Human recombit protein fragment corresponding to amino acids 1-210 of human RBFOX1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1F7 | |
| Unconjugated | |
| RBFOX1 | |
| 2BP1, A2BP1, FOX-1, FOX1, HRNBP1 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54715 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction