missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Antibody (1H3B2) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-61848
This item is not returnable.
View return policy
Description
RBFOX3/NeuN Monoclonal specifically detects RBFOX3/NeuN in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications
| RBFOX3/NeuN | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:500-1:2000, Flow Cytometry 1:200-1:400, ELISA 1:10000, Immunocytochemistry/Immunofluorescence 1:200-1:1000 | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3 | |
| Mouse | |
| 28.3 kDa | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence | |
| 1H3B2 | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| RBFOX3 | |
| Purified recombinant fragment of human RBFOX3 (AA: 1-140) expressed in E. Coli. | |
| Protein G purified | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| 146713 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction