missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Antibody (CL11896), Novus Biologicals™
Mouse Monoclonal Antibody
€ 450.00 - € 648.00
Specifications
| Antigen | RBFOX3/NeuN |
|---|---|
| Clone | CL11896 |
| Dilution | Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18601534
|
Novus Biologicals
NBP3-21203-25ul |
25 μg |
€ 450.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18671254
|
Novus Biologicals
NBP3-21203-100ul |
100 μg |
€ 648.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)Specifications
| RBFOX3/NeuN | |
| Immunohistochemistry 1:1000 - 1:2500, Immunohistochemistry-Paraffin 1:1000 - 1:2500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3 | |
| This antibody was developed against a Synthetic Peptide of A6NFN3. Actual immunogen sequence is proprietary. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| CL11896 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| PBS, pH 7.2, 40% glycerol | |
| 146713 | |
| IgG2b | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title