missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Antibody, DyLight 680, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-92716FR
This item is not returnable.
View return policy
Description
RBFOX3/NeuN Polyclonal specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating.
Specifications
| RBFOX3/NeuN | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Immunohistochemistry Free-Floating | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuronal nuclei, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Frozen) | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| RBFOX3 | |
| The N-terminal 100 amino acids of human RBFOX3/NeuN as expressed in and purified from E. coli. [UniProt# A6NFN3] | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction