missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBFOX3/NeuN Antibody (rNEUN/8055), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24074JF646
This item is not returnable.
View return policy
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| RBFOX3/NeuN | |
| Monoclonal | |
| Janelia Fluor 646 | |
| FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3 | |
| Recombinant fragment (around aa1-200) of human RBFOX3/NeuN protein (exact sequence is proprietary) | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience | |
| 146713 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Immunohistochemistry (Paraffin) | |
| rNEUN/8055 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction