Learn More
RBFOX3/NeuN Antibody (S09-7H2), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19714-100ul
 View more versions of this product
Product Code. 18675243
  • € 645.00 / 100µL
Description
RBFOX3/NeuN Monoclonal antibody specifically detects RBFOX3/NeuN in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence, Proximity Ligation Assay, Sandwich ELISA
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RBFOX3/NeuN
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot, ELISA, Immunofluorescence, Proximity Ligation Assay, Sandwich ELISA
S09-7H2
Unconjugated
FLJ56884, FLJ58356, Fox-1 homolog C, FOX3, FOX-3, hexaribonucleotide binding protein 3, HRNBP3, NeuN, neuN antigen, neuronal nuclei, neuronal nuclei antigen, RBFOX3, RNA binding protein fox-1 homolog 3, RNA binding protein, fox-1 homolog (C. elegans) 3, RNA binding protein, fox-1 homolog 3
A synthetic peptide of human RBFOX3/NeuN (Uniprot # A6NFN3)
100 μg
DNA replication Transcription Translation and Splicing, Neuronal Cell Markers, Neuroscience
146713
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.