missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBKS Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97079H
This item is not returnable.
View return policy
Description
RBKS Polyclonal specifically detects RBKS in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RBKS | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HRP | |
| EC 2.7.1.15, RBSKDKFZp686G13268,5230400M11Rik, ribokinase | |
| Produced in rabbits immunized with purified, recombinant RBKS (NP_071411.1; Met1-Phe322). | |
| 0.1 mL | |
| metabolism | |
| 64080 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction