missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM12B Antibody, Janelia Fluor™ 646, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-18499JF646
This item is not returnable.
View return policy
Description
RBM12B Polyclonal antibody specifically detects RBM12B in Human samples. It is validated for Western Blot
Specifications
| RBM12B | |
| Polyclonal | |
| Western Blot | |
| MGC:33837, RNA binding motif protein 12B, RNA-binding motif protein 12B, RNA-binding protein 12B | |
| Recognizes region between residues Between between 600 and 650.. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 389677 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction