missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM14 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2884AF647
This item is not returnable.
View return policy
Description
RBM14 Polyclonal antibody specifically detects RBM14 in Human samples. It is validated for Western Blot
Specifications
| RBM14 | |
| Polyclonal | |
| Western Blot | |
| COAAMGC31756, Paraspeckle protein 2, PSP2DKFZp779J0927, RNA binding motif protein 14, RNA-binding motif protein 14, RNA-binding protein 14, RRM-containing coactivator activator/modulator, SIPMGC15912, Synaptotagmin-interacting protein, SYT-interacting protein, SYTIP1, TMEM137, transmembrane protein 137 | |
| The immunogen recognized by this antibody maps to a region between residue 550 and 600 of human CoActivator Activator (RNA Binding Motif 14) using the numbering given in entry NP_006319.1 (GeneID 10432 ). | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 10432 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| Alexa Fluor 647 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu