missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-20102
This item is not returnable.
View return policy
Description
RBM14 Polyclonal specifically detects RBM14 in Human samples. It is validated for Western Blot.
Specifications
| RBM14 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal | |
| COAAMGC31756, Paraspeckle protein 2, PSP2DKFZp779J0927, RNA binding motif protein 14, RNA-binding motif protein 14, RNA-binding protein 14, RRM-containing coactivator activator/modulator, SIPMGC15912, Synaptotagmin-interacting protein, SYT-interacting protein, SYTIP1, TMEM137, transmembrane protein 137 | |
| Rabbit | |
| 69 kDa | |
| 0.1 mL | |
| Cell Cycle and Replication, DNA Repair | |
| 10432 | |
| Human | |
| IgG |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| 1mg/mL | |
| Western Blot 1:1000-1:10000, Immunocytochemistry/Immunofluorescence | |
| Q96PK6 | |
| RBM14 | |
| Recombinant protein encompassing a sequence within the N-terminus region of human RBM14. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Species reactivity based on sequence identity: Chimpanzee (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction