missing translation for 'onlineSavingsMsg'
Learn More

RBM15 Antibody (6B1) [DyLight 405], Novus Biologicals Biologicals™

Product Code. 30521772 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30521772 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30521772 Supplier Novus Biologicals Supplier No. NBP343026V

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

RBM15 Monoclonal antibody specifically detects RBM15 in Human samples. It is validated for ELISA,Immunohistochemistry,Immunoprecipitation,Western Blot,Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBM15
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot, Immunocytochemistry/Immunofluorescence
Classification Monoclonal
Clone 6B1
Conjugate DyLight 405
Formulation 50mM Sodium Borate
Gene Alias FLJ12479, one twenty two protein, one twenty-two, One-twenty two protein 1, putative RNA-binding protein 15, RNA binding motif protein 15, RNA-binding motif protein 15, SPEN
Host Species Mouse
Immunogen RNA binding motif protein 15 - (Uniprot# Q96T37)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64783
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.