missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM15, Mouse anti-Human, Clone: 2C1, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00064783-M19.100ug
This item is not returnable.
View return policy
Description
Sequence: PIRDRRGSLEKSQGDKRDRKNSASAERDRKHRTTAPTEGKSPLKKEDRSDGSAPSTSTASSKLKSPSQKQDGGTAPVASASPKLCLAWQGMLLLKNSNFPSNMHLLQGDLSpecifications
| RBM15 | |
| Monoclonal | |
| Unconjugated | |
| RNA binding motif protein 15 | |
| FLJ12479/FLJ21943/MGC119584/OTT/OTT1/SPEN | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA | |
| 2C1 | |
| In 1x PBS, pH 7.4 | |
| NM_022768 | |
| RBM15 | |
| RBM15 (NP_073605.4, 701 a.a. ∼ 810 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 64783 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction