missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM17 Antibody, DyLight 650, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97616C
This item is not returnable.
View return policy
Description
RBM17 Polyclonal specifically detects RBM17 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| RBM17 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 84991 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 650 | |
| MGC14439, RNA binding motif protein 17, RNA-binding motif protein 17, SPF45DKFZp686F13131, splicing factor 45,45 kDa-splicing factor, splicing factor 45kDa | |
| Produced in rabbits immunized with E. coli-derived Human RBM17 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction