missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
RBM20 Polyclonal specifically detects RBM20 in Human, Mouse samples. It is validated for Western Blot, Peptide ELISA.
Specifications
Specifications
| Dilution | Western Blot 0.3-1 ug/ml, Peptide ELISA Detection limit 1:164000 |
| Immunogen | Peptide with sequence C-QLDKAELDERP corresponding to internal region according to NP_001127835.1. |
| Purification Method | Affinity Purified |
| Quantity | 0.1 mg |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?