Learn More
RBM22 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 22103-1-AP-20UL
 View more versions of this product
Product Code. 16861645
  • € 145.00 / 20µL
Description
RanGAP1, is a homodimeric 65-kD polypeptide that specifically induces the GTPase activity of RAN, but not of RAS by over 1, 000-fold. RanGAP1 is the immediate antagonist of RCC1, a regulator molecule that keeps RAN in the active, GTP-bound state. The RANGAP1 gene encodes a 587-amino acid polypeptide. The sequence is unrelated to that of GTPase activators for other RAS-related proteins, but is 88% identical to Fug1, the murine homolog of yeast Rna1p. RanGAP1 and RCC1 control RAN-dependent transport between the nucleus and cytoplasm. RanGAP1 is a key regulator of the RAN GTP/GDP cycle.
Specifications
Specifications
RBM22
Polyclonal
Unconjugated
RBM22
fSAP47, Pre mRNA splicing factor RBM22, RBM22, RNA binding motif protein 22, ZC3H16
Rabbit
Antigen Affinity Chromatography
RUO
307410, 55696, 66810
-20°C
Liquid
Western Blot
0.65 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q4V7D7, Q8BHS3, Q9NW64
RBM22
RBM22 Fusion Protein Ag17315
20 μL
Primary
Mouse, Human, Rat