missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM23 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-81986IR
This item is not returnable.
View return policy
Description
RBM23 Polyclonal antibody specifically detects RBM23 in Human samples. It is validated for Western Blot, ELISA
Specifications
| RBM23 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| DyLight 755 | |
| CAPERbeta, FLJ10482, MGC4458, probable RNA-binding protein 23, RNA binding motif protein 23, RNA-binding motif protein 23, RNA-binding region (RNP1, RRM) containing 4, RNA-binding region-containing protein 4, RNPC4, Splicing factor SF2 | |
| RBM23 antibody was raised against a peptide corresponding to 14 amino acids near the amino terminus of human RBM23. Amino Acid Squence: YRYGHSKSPHFREKSPVR | |
| 0.1 mL | |
| Cancer, Cell Cycle and Replication | |
| 55147 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction