missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM25 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
Marca: Novus Biologicals NB100-57505
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
RBM25 Polyclonal specifically detects RBM25 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Especificaciones
| RBM25 | |
| Polyclonal | |
| Unconjugated | |
| TBS and 0.1% BSA with 0.09% Sodium Azide | |
| Arg/Glu/Asp-rich protein of 120 kDa, Arg/Glu/Asp-rich protein, 120 kDa, fSAP94, functional spliceosome-associated protein 94, MGC105088, MGC117168, NET52, Protein S164, RED120RNA-binding region-containing protein 7, RNA binding motif protein 25, RNA-binding motif protein 25, RNA-binding protein 25, RNA-binding region (RNP1, RRM) containing 7, RNPC7, S164, Snu71, U1 small nuclear ribonucleoprotein 1SNRP homolog | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 58517 | |
| Human | |
| IgG |
| Western Blot, Immunoprecipitation | |
| 0.2 mg/mL | |
| Western Blot 1:2000-1:10000, Immunoprecipitation 2-5 ug/mg lysate | |
| P49756 | |
| RBM25 | |
| The immunogen recognized by this antibody maps to a region between residue 150 and 200 of human RNA binding motif protein 25 (Arg/Glu/Asp-rich protein, 120 kDa) using the numbering given in entry AAF19255.1 (GeneID 58517). | |
| 0.1 mL | |
| Primary | |
| Based on 100% sequence identity, this antibody is predicted to react with Panda, Rhesus Monkey, Gorilla, Chimpanzee, Duckbill platypus, Tasmanian devil, White-tufted-ear marmoset and Northern white-cheeked gibbon. | |
| Store at 4C. Do not freeze. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido