Learn More
RBM25 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25297-1-AP-20UL
This item is not returnable.
View return policy
Description
RNA-binding protein that acts as a regulator of alternative pre-mRNA splicing. Involved in apoptotic cell death through the regulation of the apoptotic factor BCL2L1 isoform expression. Modulates the ratio of proapoptotic BCL2L1 isoform S to antiapoptotic BCL2L1 isoform L mRNA expression. When overexpressed, stimulates proapoptotic BCL2L1 isoform S 5′-splice site (5′-ss) selection, whereas its depletion caused the accumulation of antiapoptotic BCL2L1 isoform L. Promotes BCL2L1 isoform S 5′-ss usage through the 5′-CGGGCA-3′ RNA sequence. Its association with LUC7L3 promotes U1 snRNP binding to a weak 5′ ss in a 5′-CGGGCA-3′-dependent manner. Binds to the exonic splicing enhancer 5′-CGGGCA-3′ RNA sequence located within exon 2 of the BCL2L1 pre-mRNA. Also involved in the generation of an abnormal and truncated splice form of SCN5A in heart failure.Specifications
| RBM25 | |
| Polyclonal | |
| Unconjugated | |
| RBM25 | |
| fSAP94, NET52, Protein S164, RBM25, RED120, RNA binding motif protein 25, RNA binding protein 25, RNPC7, S164 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 58517, 67039 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.25 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| B2RY56, P49756 | |
| RBM25 | |
| RBM25 Fusion Protein Ag18244 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |