missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM26 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA575975
This item is not returnable.
View return policy
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
RBM26 (RNA Binding Motif Protein 26) is a Protein Coding gene. Diseases associated with RBM26 include Cutaneous T Cell Lymphoma and Helsmoortel-Van Der Aa Syndrome. Gene Ontology (GO) annotations related to this gene include nucleic acid binding and nucleotide binding. An important paralog of this gene is RBM27. [GeneCards].
Specifications
| RBM26 | |
| Polyclonal | |
| Unconjugated | |
| RBM26 | |
| 1700009P03Rik; acidic rich RS domain containing 2; AI447318; ARRS2; C13orf10; C230097K14Rik; C86225; CTCL tumor antigen se70-2; cutaneous T-cell lymphoma tumor antigen se70-2; fc66d05; MLZ-393; PPP1R132; Pro1777; protein expressed in male leptotene and zygotene spermatocytes 393; protein phosphatase 1, regulatory 132; Rbm26; RGD1308297; RNA binding motif protein 26; RNA-binding motif protein 26; RNA-binding protein 26; RP11-255E21.1; SE70-2; wu:fc66d05; ZC3H17; zgc:109736 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 306137, 64062, 74213 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q5T8P6, Q6NZN0 | |
| RBM26 | |
| Synthetic peptide corresponding to amino acids 726-770 of Human RBM26. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction