missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM27 Antibody, FITC, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-60426F
This item is not returnable.
View return policy
Description
RBM27 Polyclonal antibody specifically detects RBM27 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RBM27 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| RNA-binding protein 27, ARRS1, Psc1, RNA binding motif protein 27 | |
| The immunogen recognized by this antibody maps to a region between residue 700 and 750 of human RNA binding motif protein 27 using the numbering given in entry Q9P2N5.2 (GeneID 54439). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| FITC | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 54439 | |
| Store at 4C in the dark. | |
| IgG |
Correction du contenu d'un produit
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.
Nom du produit
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu