missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM28 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-12575-50ul
This item is not returnable.
View return policy
Description
RBM28 Polyclonal antibody specifically detects RBM28 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| RBM28 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry, Immunohistochemistry-Paraffin 1:50-1:200 | |
| FLJ10377, RNA binding motif protein 28, RNA-binding motif protein 28,2810480G15Rik, RNA-binding protein 28 | |
| Produced in rabbits immunized with E. coli-derived Human RBM28 fragment | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 55131 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction