missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM3 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97785PCP
This item is not returnable.
View return policy
Description
RBM3 Polyclonal antibody specifically detects RBM3 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence
Specifications
| RBM3 | |
| Polyclonal | |
| Immunocytochemistry/ Immunofluorescence | |
| IS1-RNPL, putative RNA-binding protein 3, RNA binding motif (RNP1, RRM) protein 3, RNA binding motif protein 3, RNA-binding motif protein 3, RNPL | |
| Produced in rabbits immunized with E. coli-derived Human RBM3 fragment. | |
| 0.1 mL | |
| metabolism | |
| 5935 | |
| Store at 4C in the dark. | |
| IgG |
| Immunocytochemistry | |
| PerCP | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction