missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ RBM3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551976
This item is not returnable.
View return policy
Description
Immunogen sequence: DEQALEDHFS SFGPISEVVV VKDRETQRSR GFGFITFTNP EHASVAMRAM NGESLDGRQI RVDHAGKSAR GTRGGGFGAH GRGRSYSRGG GDQGYGSGRY YDSRPGGYGY GYGRSRDYNG RNQGGYDRYS GGNY Highest antigen sequence identity to the following orthologs: Mouse - 94%, Rat - 96%.
RBM3 is a member of the glycine-rich RNA-binding protein family and a protein with one RNA recognition motif (RRM) domain. Expression of this protein is induced by cold shock and low oxygen tension.
Specifications
| RBM3 | |
| Polyclonal | |
| Unconjugated | |
| RBM3 | |
| 2600016C11Rik; hypothetical protein LOC509771; IS1-RNPL; OTTHUMP00000025800; OTTHUMP00000025802; putative RNA-binding protein 3; Rbm3; RNA binding motif (RNP1, RRM) protein 3; RNA binding motif protein 3; RNA-binding motif protein 3; RNA-binding protein 3; RNPL | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114488, 19652, 5935 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O89086, P98179, Q925G0 | |
| RBM3 | |
| Recombinant protein corresponding to Human RBM3. Recombinant protein control fragment (Product #RP-92189). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction