missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM33 Antibody, PE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-22280PE
This item is not returnable.
View return policy
Description
RBM33 Polyclonal antibody specifically detects RBM33 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation (Negative)
Specifications
| RBM33 | |
| Polyclonal | |
| Western Blot, Immunoprecipitation (Negative) | |
| DKFZp434D1319, MGC20460, proline-rich protein 8, RNA binding motif protein 33, RNA-binding motif protein 33, RNA-binding protein 33 | |
| The immunogen this antibody was made to, maps to a region between residue 500 and 550 of human RNA Binding Motif Protein 33 using the numbering given in entry NP_444271.2 (GeneID 155435). | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot | |
| PE | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 155435 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction