missing translation for 'onlineSavingsMsg'
Learn More
Learn More
RBM34 Antibody, Alexa Fluor™ 647, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97638AF647
This item is not returnable.
View return policy
Description
RBM34 Polyclonal specifically detects RBM34 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| RBM34 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 23029 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Alexa Fluor 647 | |
| KIAA0117RNA-binding motif protein 34, RNA binding motif protein 34, RNA-binding protein 34 | |
| Produced in rabbits immunized with E. coli-derived Human RBM34 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction