Learn More
Invitrogen™ RBM34 Polyclonal Antibody
Description
Immunogen sequence: FSSLEPQIQP VYVPVPKQTI KKTKRNEEEE STSQIERPLS QEPAKKVKAK KKHTNAEKKL ADRESALASA DLEEEIHQKQ GQKRKNSQPG VKVADRKILD DTEDTVVSQR KKIQINQEEE RLKNERTVFV GNLPVTCNKK KLKSFFKEYG QIESVRFRSL IPAEGTLSKK LAAIKRKIHP DQKNINAYVV FKEESAATQA LKRNGAQIAD GFRIRVDLAS E; Positive Samples: HeLa, Jurkat, A-549, 293T, K-562; Cellular Location: Nucleus, nucleolus.
Specifications
Specifications
| Antigen | RBM34 |
| Applications | ELISA, Western Blot |
| Classification | Polyclonal |
| Concentration | 0.89 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RBM34 |
| Gene Accession No. | P42696 |
| Gene Alias | 4930547K05Rik; 6330444B11; D8Ertd233e; KIAA0117; Rbm34; RGD1310161; RNA binding motif protein 34; RNA-binding motif protein 34; RNA-binding protein 34 |
| Gene Symbols | RBM34 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.